Learn More
Biomatik Corporation Human Elafin (Pi3) Elisa Kit
Supplier:  Biomatik Corporation EKC42597
Encompass_Preferred
Human Elafin (Pi3) Elisa Kit for detecting Peptidase Inhibitor 3, Skin-Derived in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19957; PI3. Target Synonyms: Cementoin; ELAF_HUMAN; Elafin; Elafin/Skalp; Elastase Specific Inhibitor; Elastase-specific inhibitor; ES 1; ESI; Peptidase inhibitor 3; Peptidase inhibitor 3; skin derived; PI 3; PI-3; PI3; Pre elafin; Protease inhibitor 3 skin derived; Protease inhibitor 3; skin derived (SKALP); Protease inhibitor WAP3; SKALP; Skin derived Anti leukoproteinase; Skin-derived antileukoproteinase; Trappin 2; WAP four disulfide core domain 14; WAP four disulfide core domain protein 14; WAP four-disulfide core domain protein 14; WAP3; WFDC14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5136