Learn More
Biomatik Corporation Rat Regenerating Islet-Derived Protein 3-Gamma (Reg3G) Elisa Kit
Supplier:  Biomatik Corporation EKC42604
Encompass_Preferred
Rat Regenerating Islet-Derived Protein 3-Gamma (Reg3G) Elisa Kit for detecting Regenerating Islet-Derived 3 Gamma in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P42854; REG3G. Target Synonyms: Reg3g; Pap3; Regenerating islet-derived protein 3-gamma; REG-3-gamma; Pancreatitis-associated protein 3; Regenerating islet-derived protein III-gamma; Reg III-gamma) [Cleaved into: Regenerating islet-derived protein 3-gamma 16.5 kDa form; Regenerating islet-derived protein 3-gamma 15 kDa form]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5139