Learn More
Biomatik Corporation Mouse Secretoglobin family 3A member 1(SCGB3A1) ELISA kit
Supplier:  Biomatik Corporation EKC42611
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 pg/ml -4000 pg/ml. Sensitivity: 15.6 pg/ml. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q920D7. Target Synonyms: HIN-1, HIN1, LU105, MGC87867, PnSP-2, UGRP2, cytokine high in normal-1|pneumo secretory protein 2. Restriction: For Research Use Only.
Catalog No. 50-229-5141