Learn More
Biomatik Corporation Rat Sclerostin(SOST) ELISA kit
Supplier:  Biomatik Corporation EKC42645
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 18.75 pg/mL-1200 pg/mL. Sensitivity: 4.68 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q99P67. Target Synonyms: VBCH, sclerostin. Restriction: For Research Use Only.
Catalog No. 50-229-5154