Learn More
Biomatik Corporation Human Secreted Phosphoprotein 2 (Spp2/Spp24) Elisa Kit
Supplier:  Biomatik Corporation EKC42651
Encompass_Preferred
Human Secreted Phosphoprotein 2 (Spp2/Spp24) Elisa Kit for detecting Secreted Phosphoprotein 2, 24Kda in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13103; SPP2. Target Synonyms: SPP2; SPP24; Secreted phosphoprotein 24; Spp-24; Secreted phosphoprotein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5158