Learn More
Biomatik Corporation Rat ACTα2 (Actin Alpha 2, Smooth Muscle) ELISA Kit
Supplier:  Biomatik Corporation EKF58906
Encompass_Preferred
Rat ACTα2 (Actin Alpha 2, Smooth Muscle) ELISA Kit for detecting ACTα2 in cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P62738. Target Synonyms: ACTα2; Actin Alpha 2; Smooth Muscle; ACTA2; AAT6; ACTSA; MYMY5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6145