Learn More
Biomatik Corporation Human DEFB129 (Beta-defensin 129) ELISA Kit
Supplier:  Biomatik Corporation EKF58916
Encompass_Preferred
Human DEFB129 (Beta-defensin 129) ELISA Kit for detecting DEFB129 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9H1M3. Target Synonyms: DEFB129; Beta-defensin 129; Beta-defensin 29; DEFB-29; Defensin; beta 129. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6154