Learn More
Biomatik Corporation Human BTN2A1 (Butyrophilin subfamily 2 member A1) ELISA Kit
Supplier:  Biomatik Corporation EKF58922
Encompass_Preferred
Human BTN2A1 (Butyrophilin subfamily 2 member A1) ELISA Kit for detecting BTN2A1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7KYR7. Target Synonyms: BTN2A1; Butyrophilin subfamily 2 member A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6157