Learn More
Biomatik Corporation Rat BACE1 (Beta Site APP Cleaving Enzyme 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58924
Encompass_Preferred
Rat BACE1 (Beta Site APP Cleaving Enzyme 1) ELISA Kit for detecting BACE1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P56819. Target Synonyms: BACE1; APP beta-secretase; ASP2; Aspartyl protease 2; BACEAsp 2; beta-secretase 1; beta-secretase 1 precursor variant 1; beta-site amyloid beta A4 precursor protein-cleaving enzyme; Beta-site amyloid precursor protein cleaving enzyme 1; Beta-site APP cleaving enzyme 1; beta-site APP-cleaving enzyme; beta-site APP-cleaving enzyme 1; EC 3.4.23; EC 3.4.23.46; FLJ90568; HSPC104; KIAA1149; memapsin-2; Memapsin-2; Membrane-associated aspartic protease 2; transmembrane aspartic proteinase Asp2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6159