Learn More
Biomatik Corporation Mouse Sostdc1 (Sclerostin domain-containing protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58935
Encompass_Preferred
Mouse Sostdc1 (Sclerostin domain-containing protein 1) ELISA Kit for detecting Sostdc1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9CQN4. Target Synonyms: Sostdc1; CDA019; Ectodin; WISE; cystine-knot containing secreted protein; Ectodermal BMP inhibitor; USAG-1; USAG1; uterine sensitization-associated protein-1; Uterine sensitization-associated gene 1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6167