Learn More
Biomatik Corporation Human PRF1 (Perforin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58945
Encompass_Preferred
Human PRF1 (Perforin-1) ELISA Kit for detecting PRF1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14222. Target Synonyms: PRF1; Perforin-1; Cytolysin; FLH2; Lymphocyte pore-forming protein; P1; PERF_Human; Perforin 1; PFP; Pore forming protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6174