Learn More
Biomatik Corporation Human S100A8 (Protein S100-A8) ELISA Kit
Supplier:  Biomatik Corporation EKF58954
Encompass_Preferred
Human S100A8 (Protein S100-A8) ELISA Kit for detecting S100A8 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05109. Target Synonyms: S100A8; S100 Calcium Binding Protein A8; Calgranulin A; CFAG; MRP8; CAGACP-10; Calgranulin-A; Calprotectin L1L subunit; CFAGL1Ag; CGLA; Cystic fibrosis antigen; Leukocyte L1 complex light chain; MigRation inhibitory factor-related protein 8; MRP-8; NIF; P8; protein S100-A8; S100 calcium binding protein A8; Urinary stone protein band A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6181