Learn More
Biomatik Corporation Human PLK1 (Serine/threonine-protein kinase PLK1) ELISA Kit
Supplier:  Biomatik Corporation EKF58960
Encompass_Preferred
Human PLK1 (Serine/threonine-protein kinase PLK1) ELISA Kit for detecting PLK1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P53350. Target Synonyms: PLK1; Serine; threonine-protein kinase PLK1; Polo-like kinase 1; PLK-1; Serine; threonine-protein kinase 13; STPK13; PLK; cell cycle regulated protein kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6185