Learn More
Biomatik Corporation Human HSD3B1 (3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58963
Encompass_Preferred
Human HSD3B1 (3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1) ELISA Kit for detecting HSD3B1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P14060. Target Synonyms: HSD3B1; 3 beta-hydroxysteroid dehydrogenase; Delta 5-->4-isomerase type I; 3-beta-HSD I; Trophoblast antigen FDO161G; 3BH; HSDB3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6188