Learn More
Biomatik Corporation Mouse ANGPTL2 (Angiopoietin Like Protein 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58974
Encompass_Preferred
Mouse ANGPTL2 (Angiopoietin Like Protein 2) ELISA Kit for detecting ANGPTL2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9R045. Target Synonyms: ANGPTL2; ANGRP2; angiopoietin-like 2; Angiopoietin-like protein 2; angiopoietin-related protein 2; ARP2HARP;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6197