Learn More
Biomatik Corporation Mouse GKN2 (Gastrokine 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58981
Encompass_Preferred
Mouse GKN2 (Gastrokine 2) ELISA Kit for detecting GKN2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9CQS6. Target Synonyms: Gastrokine-2; Blottin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6202