Learn More
Biomatik Corporation Rat CASP9 (Caspase 9) ELISA Kit
Supplier:  Biomatik Corporation EKF58986
Encompass_Preferred
Rat CASP9 (Caspase 9) ELISA Kit for detecting CASP9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q920G4. Target Synonyms: CASP9; Caspase 9; Apoptotic protease Mch 6; p35; p10; Cas9; APAF-3; APAF3; apoptotic protease activating factor 3; Apoptotic protease Mch-6; Apoptotic protease-activating factor 3; CASP-9; apoptosis-related cysteine peptidase; caspase-9; ICE-LAP6; CASPASE-9c; ICE-like apoptotic protease 6; MCH6apoptosis-related cysteine protease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6206