Learn More
Biomatik Corporation Human COMT (Catechol O-methyltransferase) ELISA Kit
Supplier:  Biomatik Corporation EKF58998
Encompass_Preferred
Human COMT (Catechol O-methyltransferase) ELISA Kit for detecting COMT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21964. Target Synonyms: COMT; catechol O-methyltransferase; catechol-O-methyltransferase; EC 2.1.1.6;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6217