Learn More
Biomatik Corporation Human PRAP1 (Proline-rich acidic protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF59011
Encompass_Preferred
Human PRAP1 (Proline-rich acidic protein 1) ELISA Kit for detecting PRAP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-200pg/ml. Sensitivity: 1.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q96NZ9. Target Synonyms: PRAP1; Proline-rich acidic protein 1; Uterine-specific proline-rich acidic protein; Uterine-specific proline-rich acidic protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6228