Learn More
Biomatik Corporation Mouse Dnase1l3 (Deoxyribonuclease gamma) ELISA Kit
Supplier:  Biomatik Corporation EKF59044
Encompass_Preferred
Mouse Dnase1l3 (Deoxyribonuclease gamma) ELISA Kit for detecting Dnase1l3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O55070. Target Synonyms: Dnase1l3; Deoxyribonuclease gamma; DNase gamma; Liver and spleen Dnase; LS-DNase; LSD; DNase I homolog protein DHP2; Deoxyribonuclease I-like 3; DNase I-like 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6253