Learn More
Biomatik Corporation Rat MyD88 (myeloid differentiation factor 88) ELISA Kit
Supplier:  Biomatik Corporation EKF59052
Encompass_Preferred
Rat MyD88 (myeloid differentiation factor 88) ELISA Kit for detecting MyD88 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q6Y1S1. Target Synonyms: MYD88; MYD88D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6260