Learn More
Biomatik Corporation Human HMGCR (3-hydroxy-3-methylglutaryl-coenzyme A reductase) ELISA Kit
Supplier:  Biomatik Corporation EKF59055
Encompass_Preferred
Human HMGCR (3-hydroxy-3-methylglutaryl-coenzyme A reductase) ELISA Kit for detecting HMGCR in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04035. Target Synonyms: HMGCR; 3-hydroxy-3-methylglutaryl-coenzyme A reductase; HMG-CoA reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6262