Learn More
Biomatik Corporation Human LPCAT2 (Lysophosphatidylcholine acyltransferase 2) ELISA Kit
Supplier:  Biomatik Corporation EKF59071
Encompass_Preferred
Human LPCAT2 (Lysophosphatidylcholine acyltransferase 2) ELISA Kit for detecting LPCAT2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7L5N7. Target Synonyms: LPCAT2; LPC acyltransferase 2; LPCAT-2; LysoPC acyltransferase 2; Lysophosphatidic acid acyltransferase alpha; LPAAT-alpha; Acetyl-CoA; lyso-PAF acetyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6276