Learn More
Biomatik Corporation Mouse DHEA-S (Dehydroepiandrosterone sulfate) ELISA Kit
Supplier:  Biomatik Corporation EKF59078
Encompass_Preferred
Mouse DHEA-S (Dehydroepiandrosterone sulfate) ELISA Kit for detecting DHEA-S in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000ng/ml. Sensitivity: 9.375ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: DHEA-S; DHEAS; DHEA-ST; hSTa; ST2A3; SULT2A1; alcohol; hydroxysteroid sulfotransferase; Dehydroepiandrosterone sulfotransferase; DHEAS; DHEA-STbile salt sulfotransferase; EC 2.8.2.14; HST; hSTa; Hydroxysteroid Sulfotransferase; ST2; ST2A1; ST2A3; STDbile-salt sulfotranasferase 2A1; Sulfotransferase 2A1; sulfotransferase family; cytosolic; 2A; dehydroepiandrosterone; DHEA-preferring; member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6283