Learn More
Biomatik Corporation Human KLB (Beta-klotho) ELISA Kit
Supplier:  Biomatik Corporation EKF59086
Encompass_Preferred
Human KLB (Beta-klotho) ELISA Kit for detecting KLB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q86Z14. Target Synonyms: KLB; BKL; Beta-klotho; BetaKlotho; Klotho beta-like protein; klotho beta; klotho beta like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6288