Learn More
Biomatik Corporation Human ADAMTS13 (A disintegrin and metalloproteinase with thrombospondin motifs 13) ELISA Kit
Supplier:  Biomatik Corporation EKF59091
Encompass_Preferred
Human ADAMTS13 (A disintegrin and metalloproteinase with thrombospondin motifs 13) ELISA Kit for detecting ADAMTS13 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20miu/ml. Sensitivity: 0.188miu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q76LX8. Target Synonyms: ADAMTS13; VWFCP; TTP; vWF-CP; a disintegrin-like and metalloprotease; reprolysin type with thrombospondintype 1 motif; 13; ADAM metallopeptidase with thrombospondin type 1 motif; 13; ADAM-TS13; ADAMTS-13; vWF-cleaving protease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6293