Learn More
Biomatik Corporation Mouse TSP-1 (Thrombin Sensitive Protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF59098
Encompass_Preferred
Mouse TSP-1 (Thrombin Sensitive Protein 1) ELISA Kit for detecting TSP-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35441. Target Synonyms: TSP-1; Thrombospondin-1; THBS1; TSP-1; THBS; THBS-1; thrombospondin 1; thrombospondin-1p180; TSP1thrombospondin-1; TSPTSP-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6299