Learn More
Biomatik Corporation Human LAMB3 (Laminin subunit beta-3) ELISA Kit
Supplier:  Biomatik Corporation EKF59109
Encompass_Preferred
Human LAMB3 (Laminin subunit beta-3) ELISA Kit for detecting LAMB3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13751. Target Synonyms: LAMB3; Laminin subunit beta-3; Nicein subunit beta; Laminin-5 subunit beta; Laminin B1k chain; Kalinin subunit beta; Epiligrin subunit bata; Kalinin B1 chain; LAM5; LAMNB1; BM600-125KDA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6309