Learn More
Biomatik Corporation Human RUVBL1 (RuvB-like 1) ELISA Kit
Supplier:  Biomatik Corporation EKF59127
Encompass_Preferred
Human RUVBL1 (RuvB-like 1) ELISA Kit for detecting RUVBL1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9Y265. Target Synonyms: RUVBL1; TIP49a; Pontin 52; Nuclear matrix protein 238; NMP 238; INO80 complex subunit H; TIP60-associated protein 54-alpha; TAP54-alpha; 49 kDa TATA box-binding protein-interacting protein; 49 kDa TBP-interacting protein; 54 kDa erythrocyte cytosolic protein; ECP-54;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6324