Learn More
Biomatik Corporation Mouse NOX4 (Nicotinamide Adenine Dinucleotide Phosphate Oxidase 4) ELISA Kit
Supplier:  Biomatik Corporation EKF59128
Encompass_Preferred
Mouse NOX4 (Nicotinamide Adenine Dinucleotide Phosphate Oxidase 4) ELISA Kit for detecting NOX4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9JHI8. Target Synonyms: NOX4; KOX-1; RENOX; EC 1.6.3; Kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX; KOX-1Renal NAD; PH-oxidase; NADPH oxidase 4; Superoxide-geneRating NADPH oxidase 4; Renal NAD; PH-oxidase; Kidney oxidase-1; KOX-1; Kidney superoxide-producing NADPH oxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6325