Learn More
Biomatik Corporation Human LIF (Leukemia inhibitory factor) ELISA Kit
Supplier:  Biomatik Corporation EKF59129
Encompass_Preferred
Human LIF (Leukemia inhibitory factor) ELISA Kit for detecting LIF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15018. Target Synonyms: LIF; Leukemia Inhibitory Factor; CDF; D factor; DIA; differentiation inhibitory activity; differentiation stimulating factor; Differentiation-stimulating factor; Emfilermin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6326