Learn More
Biomatik Corporation Human HTR1B (5-hydroxytryptamine receptor 1B) ELISA Kit
Supplier:  Biomatik Corporation EKF59135
Encompass_Preferred
Human HTR1B (5-hydroxytryptamine receptor 1B) ELISA Kit for detecting HTR1B in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P28222. Target Synonyms: HTR1B; 5-HT1B; 5-hydroxytryptamine receptor 1B; 5HT1DB; HTR1D2; HTR1DB; S12; Serotonin 1D beta receptor; Serotonin receptor 1B; 5-HT-1B; 5-HT1DB; 5-HT-1D-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6332