Learn More
Biomatik Corporation Human C3b (Complement Fragment 3b) ELISA Kit
Supplier:  Biomatik Corporation EKF59149
Encompass_Preferred
Human C3b (Complement Fragment 3b) ELISA Kit for detecting C3b in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01024. Target Synonyms: C3b. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6344