Learn More
Biomatik Corporation Rat Mat1a (S-adenosylmethionine synthase isoform type-1) ELISA Kit
Supplier:  Biomatik Corporation EKF59151
Encompass_Preferred
Rat Mat1a (S-adenosylmethionine synthase isoform type-1) ELISA Kit for detecting Mat1a in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13444. Target Synonyms: Mat1a; AMS1; MATA1; AdoMet synthase 1; S-adenosylmethionine synthase isoform type-1; Methionine adenosyltransferase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6346