Learn More
Biomatik Corporation Human MAPK14 (Mitogen-activated protein kinase 14) ELISA Kit
Supplier:  Biomatik Corporation EKF59164
Encompass_Preferred
Human MAPK14 (Mitogen-activated protein kinase 14) ELISA Kit for detecting MAPK14 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q16539. Target Synonyms: MAPK14; SAPK2A; SAPK2a; Mitogen-activated protein kinase p38 alpha; MAP kinase p38 alpha; MAX-interacting protein 2; Cytokine suppressive anti-inflammatory drug-binding protein; CSAID-binding protein; CSBP; MAP kinase MXI2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6356