Learn More
Biomatik Corporation Mouse Lrp1 (Prolow-density lipoprotein receptor-related protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF59166
Encompass_Preferred
Mouse Lrp1 (Prolow-density lipoprotein receptor-related protein 1) ELISA Kit for detecting Lrp1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q91ZX7. Target Synonyms: Lrp1; LRP1; Prolow-density lipoprotein receptor-related protein 1; LRP-1; Alpha-2-macroglobulin receptor; A2MR; Apolipoprotein E receptor; APOER; CD91. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6358