Learn More
Biomatik Corporation Rat Ldhb (L-lactate dehydrogenase B chain) ELISA Kit
Supplier:  Biomatik Corporation EKF59186
Encompass_Preferred
Rat Ldhb (L-lactate dehydrogenase B chain) ELISA Kit for detecting Ldhb in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P42123. Target Synonyms: Ldhb; LDH-B; LDH-H. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6373