Learn More
Biomatik Corporation Mouse FMOD (Fibromodulin) ELISA Kit
Supplier:  Biomatik Corporation EKF59188
Encompass_Preferred
Mouse FMOD (Fibromodulin) ELISA Kit for detecting FMOD in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100pg/ml. Sensitivity: 0.938pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P50608. Target Synonyms: FMOD; FM; SLRR2E; Collagen-binding 59 kDa protein; fibromodulin; fibromodulin proteoglycan; FM; KeRatan sulfate proteoglycan fibromodulin; SLRR2EKSPG fibromodulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6375