Learn More
Biomatik Corporation Human PRG4 (Proteoglycan 4) ELISA Kit
Supplier:  Biomatik Corporation EKF59194
Encompass_Preferred
Human PRG4 (Proteoglycan 4) ELISA Kit for detecting PRG4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q92954. Target Synonyms: PRG4; Proteoglycan 4; SZP; Superficial zone proteoglycan; Lubricin; Megakaryocyte-stimulating factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6381