Learn More
Biomatik Corporation Mouse Comp (Cartilage oligomeric matrix protein) ELISA Kit
Supplier:  Biomatik Corporation EKF59197
Encompass_Preferred
Mouse Comp (Cartilage oligomeric matrix protein) ELISA Kit for detecting Comp in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9R0G6. Target Synonyms: Comp; Cartilage Oligomeric Matrix Protein; COMP; EDM1; EPD1; MED; PSACH; THBS5; TSP-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6384