Learn More
Biomatik Corporation Human LDL (Low Density Lipoprotein) ELISA Kit
Supplier:  Biomatik Corporation EKF59203
Encompass_Preferred
Human LDL (Low Density Lipoprotein) ELISA Kit for detecting LDL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400nmol/ml. Sensitivity: 3.75nmol/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04114. Target Synonyms: LDL; Low-density lipoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6389