Learn More
Biomatik Corporation Mouse HHIP (Hedgehog Interacting Protein) ELISA Kit
Supplier:  Biomatik Corporation EKF59206
Encompass_Preferred
Mouse HHIP (Hedgehog Interacting Protein) ELISA Kit for detecting HHIP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7TN16. Target Synonyms: HHIP; HIP; hedgehog-interacting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6391