Learn More
Biomatik Corporation Mouse β-CTx (Beta Crosslaps) ELISA Kit
Supplier:  Biomatik Corporation EKF59213
Encompass_Preferred
Mouse β-CTx (Beta Crosslaps) ELISA Kit for detecting β-CTx in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P11087; P28481. Target Synonyms: β-CTx. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6396