Learn More
Biomatik Corporation Human PAP (Plasmin-Antiplasmin Complex) ELISA Kit
Supplier:  Biomatik Corporation EKF59462
Encompass_Preferred
Human PAP (Plasmin-Antiplasmin Complex) ELISA Kit for detecting PAP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P51003. Target Synonyms: Plasmin-Antiplasmin Complex. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6409