Learn More
Biomatik Corporation Human IL-17 F (Interleukin 17F) ELISA Kit
Supplier:  Biomatik Corporation EKF59467
Encompass_Preferred
Human IL-17 F (Interleukin 17F) ELISA Kit for detecting IL-17 F in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q96PD4. Target Synonyms: IL-17 F. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6411