Learn More
Biomatik Corporation Rat IL-20 (Interleukin 20) ELISA Kit
Supplier:  Biomatik Corporation EKF59751
Encompass_Preferred
Rat IL-20 (Interleukin 20) ELISA Kit for detecting IL-20 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: B8K1Y1. Target Synonyms: IL-20; IL20; ZCYTO10; IL10D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6428