Learn More
Biomatik Corporation Rat Gpbar1 (G-protein coupled bile acid receptor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF59804
Encompass_Preferred
Rat Gpbar1 (G-protein coupled bile acid receptor 1) ELISA Kit for detecting Gpbar1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q80T02. Target Synonyms: Gpbar1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6431