Learn More
Biomatik Corporation fT3 (Free Triiodothyronine) ELISA Kit
Supplier:  Biomatik Corporation EKF59866
Encompass_Preferred
fT3 (Free Triiodothyronine) ELISA Kit for detecting fT3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 0.781-50pg/ml. Sensitivity: 0.469pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: fT3; Free Triiodothyronine Indes. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6433