Learn More
Biomatik Corporation Human IL-17RA (Interleukin 17 receptor A) ELISA Kit
Supplier:  Biomatik Corporation EKF59983
Encompass_Preferred
Human IL-17RA (Interleukin 17 receptor A) ELISA Kit for detecting IL-17RA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q96F46. Target Synonyms: IL-17RA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6442