Learn More
Biomatik Corporation Human CYGB (Cytoglobin) ELISA Kit
Supplier:  Biomatik Corporation EKF60028
Encompass_Preferred
Human CYGB (Cytoglobin) ELISA Kit for detecting CYGB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8WWM9. Target Synonyms: CYGB; cytoglobin; HGB; Histoglobin; STAP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6444